missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene oxide, M.W. 300,000, Thermo Scientific Chemicals

Catalog Number p-7051912
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO

Polyethylene oxide, M.W. 300,000 is used to impart viscosity to and modify flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
pH 5 to 7
Physical Form Powder
Melting Point 65°C
Density 1.21 g/mL
Solubility Information Soluble in acetone,alcohol,chloroform,toluene and dichloromethane.
Quantity 25 g
Odor Odorless
Refractive Index 1.4539
Chemical Name or Material Polyethylene oxide, M.W. 300,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'