Learn More
Socorex™ Polyethylene Nozzle Protection Filter
Use the Socorex™ Polyethylene Nozzle Protection Filter to guard against liquid overflow or aerosol contamination. The filter is interchangeable, non-sterile and non-autoclavable.
Brand: Socorex™ 322.810
Code : A0
Additional Details : Weight : 0.00630kg
Description
- Non-autoclavable
- Non-sterilised
- 100 Pack
- Polyethylene (PE)
Specifications
No | |
Nozzle Protection Filter | |
Acura™ manual 810 (for autoclavable Acura 810 model only) |
Polyethylene | |
Nozzle Protection | |
100Pack |