missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Graduated Wide Neck Bottles

Designed to withstand considerable pressure before rupture.

Brand:  Kartell™ 411

Code : 02

Additional Details : Weight : 0.50000kg

Product Code. 11341165

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Polyethylene body with polypropylene cap
  • Secured by tagging points on bottle and cap
  • Confidential notes can be placed in space between insert sealing plug and cap
  • Suitable for storing food stuff
Specifications

Specifications

500mL
Yes
Polyethylene
Graduated Wide Neck Bottle
50mL
EC Regulation 1935/2004, EC Regulation 10/2011
Foodstuff
34.5mm
Bottle
Wide
Videos