missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Graduated Wide Neck Bottles

Designed to withstand considerable pressure before rupture.

Brand:  Kartell™ 409

Code : 02

Additional Details : Weight : 0.52500kg

 View more versions of this product

Product Code. 11331165

Quantity Price
1 £29.00 / Pack of 10
10 £26.54 / Pack of 10
In Stock 63

Add to basket


Description

Description

  • Polyethylene body with polypropylene cap
  • Secured by tagging points on bottle and cap
  • Confidential notes can be placed in space between insert sealing plug and cap
  • Suitable for storing food stuff
Specifications

Specifications

100mL
Yes
Polyethylene
Graduated Wide Neck Bottle
20mL
EC Regulation 1935/2004, EC Regulation 10/2011
Foodstuff
24mm
Bottle
Wide
Videos