missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Graduated Wide Mouth Wash Bottles

Wide mouth design aids easy and safe filling of contents.

Brand:  Kartell™ 1638

Code : NEW

Additional Details : Weight : 0.04400kg

 View more versions of this product

Product Code. 11312884

  • £2.70 / Each


In Stock 18
Add to basket

Description

Description

  • Made from polyethylene
  • Color-coded caps for easy identification
  • Ingenious cap designed to avoid involuntary dripping
  • Suitable for storing food stuff
Specifications

Specifications

500mL
Yes
Polyethylene
GL45
100mL
Yes
38mm
Color coded cap
Bottle, Wide Mouth Wash
Red
Wide
Videos