missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Graduated Wide Mouth Wash Bottles

Wide mouth design aids easy and safe filling of contents.

Brand:  Kartell™ 1637-06

Code : 02

Additional Details : Weight : 0.20680kg

 View more versions of this product

Product Code. 12398248

Quantity Price
1 £30.45 / Pack of 5
10 £27.86 / Pack of 5
In Stock 1

Add to basket


Description

Description

  • Made from polyethylene
  • Color-coded caps for easy identification
  • Ingenious cap designed to avoid involuntary dripping
  • Suitable for storing food stuff
Specifications

Specifications

250mL
Yes
Polyethylene
GL45
25mL
Yes
38mm
Color coded cap
Bottle, Wide Mouth Wash
Yellow
Wide
Videos
Documentation

Technical Documents