missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Series 300 Graduated Narrow Neck Bottles

Feature double-sealing with insert cap to ensure tightness.

Brand:  Kartell™ 325

Code : 02

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11341235

Quantity Price
1 £31.15 / Pack of 5
10 £28.50 / Pack of 5
In Stock 36

Add to basket


Description

Description

  • Made from polyethylene
  • Sloping shoulders reduce tendency for residue deposits
  • No leaking with easy-grip cap
  • Tagging points on shoulder and cap ensure tamper-proof closure
  • Suitable for storing food stuff
Specifications

Specifications

1000mL
Yes
33mm
100mL
Insert cap
EC 1935/2004, EC 10/2011
LDPE
Bottle, Narrow Neck
Narrow
Videos
Documentation