missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Polyethylene glycol methyl ether methacrylate, M.W. 1,000

Brand:  Alfa Aesar™ 046991.MD

Code : NEW

Additional Details : CAS Number : 26915-72-0 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15407365

  • £18.40 / 250mg


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Polyethylene glycol methyl ether methacrylate
M.W. 1,000
MFCD00241432
26915-72-0
CH3(OCH2CH2)nOCOC(=CH2)CH3
mPEG-methacrylate

Specifications

Polyethylene glycol methyl ether methacrylate
26915-72-0
CH3(OCH2CH2)nOCOC(=CH2)CH3
mPEG-methacrylate
33° to 38°C
M.W. 1,000
Granular Solid
MFCD00241432
White to off-white
250mg
Videos
SDS