missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol-lithium acetate soln., Thermo Scientific™

Catalog Number p-7042343
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
PEG-LiOAc soltuion; PEG-lithium acetate solution
This item is not returnable. View return policy
PEG-LiOAc soltuion; PEG-lithium acetate solution

Common solution for protein and DNA isolation from yeast

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Polyethylene glycol-lithium acetate soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'