missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 400, Liquid, BAKER™, J.T. Baker™
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Physical Form | Liquid |
Quantity | 500 mL |
Formula Weight | 380 to 420 (Average) |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.15kg |
Chemical Name or Material | Polyethylene Glycol 400 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'