missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 400, Thermo Scientific Chemicals

Catalog Number p-7026155
Click to view available options
Quantity:
250 g
1000 g
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG 400
ethane-1,2-diol
[H]OCCO
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG 400
ethane-1,2-diol
[H]OCCO

Polyethylene glycol is used in a variety of pharmaceutical formulations. It finds application in Hewlett-Packard designjet printers as an ink solvent and a lubricant for the print heads. It is an important raw material for the manufacture of polyuretnae, PEG salts of lauric, oleic and stearic acids and latex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
pH 5 to 7
Melting Point 4°C to 8°C
Linear Formula H(OCH2CH2)nOH
Density 1.126
Merck Index 14,7568
Solubility Information Miscible with water,acetone,alcohols,benzene,glycerin,glycols and aromatic hydrocarbons. Slightly miscible with aliphatic hydrocarbons.
Quantity 250 g
Flash Point 240°C (464°F)
Odor Odorless
Refractive Index 1.466
Sensitivity Hygroscopic
Chemical Name or Material Polyethylene glycol 400
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'