missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 3350, Powder, BAKER™, J.T. Baker™
Click to view available options
Quantity:
1 kg
2 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Powder |
Quantity | 2 kg |
Formula Weight | 3000 to 3700 (Average) |
Packaging | Poly Pail |
Chemical Name or Material | Polyethylene Glycol 3350 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'