missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol 12,000, Thermo Scientific Chemicals
33.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Polyethylene glycol is used to modify therapeutic proteins and peptides to increase their solubility. In gas chromatography, it is used as a polar stationary phase. It is also useful in preserving objects obtained from the ocean. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Melting Point | 64°C to 65°C |
Merck Index | 14,7568 |
Solubility Information | Soluble in organic solvents,water and aromatic hydrocarbons. Slightly soluble in aliphatic hydrocarbons. |
Quantity | 250 g |
Flash Point | >240°C (464°F) |
Chemical Name or Material | Polyethylene Glycol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'