missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 1,000, Thermo Scientific Chemicals

Catalog Number p-3173387
Click to view available options
Quantity:
250 g
1000 g
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO

Polyethylene glycol is used as an excipent in pharmaceutical products. It is used in the precipitation of proteins as well as in the separation and purification of biomolecules and in the induction of cell hybridization. It acts as a fusing agent to enhance the effect of macrophages on hybridoma; as a vascular agent in preclinical work; as an anti-foaming agent in food and as the gate insulator in an electric double-layer transistor to enhance superconductivity in an insulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Waxy Solid
Melting Point 35°C to 40°C
Density 1.101
Solubility Information Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons.
Quantity 250 g
Flash Point 260°C (500°F)
Chemical Name or Material Polyethylene glycol 1000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'