Learn More
Macherey-Nagel™ Polyethylene Frit
£109.00 - £153.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10680152
|
Macherey-Nagel™
732019 |
LV Polypropylene Columns 15 mL, 100 mg Adsorbent Weight |
£109.00
Pack of 250 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10083141
|
Macherey-Nagel™
732020 |
LV Polypropylene Columns 15 mL, 200/500 mg Adsorbent Weight |
£153.00
Pack of 250 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Specifications
Frit | |
Polyethylene | |
250 Pack |