missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polyethylene Flat-Bottom Sample Vials

General purpose containers for laboratory weighing and storage needs.

Brand:  Kartell™ 730

Code : 02

Additional Details : Weight : 0.00752kg

 View more versions of this product

Product Code. 12315749

Quantity Price
1 £5.34 / Pack of 10
10 £4.89 / Pack of 10
In Stock 1

Add to basket


Description

Description

  • Convenient push-on attached hinged lids
  • Lightweight and unbreakable
  • Translucent
  • Suitable for storing food stuff
Specifications

Specifications

1mL
Polyethylene
Flat Bottom Sample Vial
Push on hinged lid
0.98mm
Push on hinged lid
Videos
Documentation