missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polyethylene Flat-Bottom Sample Vials

General purpose containers for laboratory weighing and storage needs.

Brand:  Kartell™ 733

Code : 02

Additional Details : Weight : 0.03290kg

Product Code. 11381215

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Convenient push-on attached hinged lids
  • Lightweight and unbreakable
  • Translucent
  • Suitable for storing food stuff
Specifications

Specifications

7mL
Polyethylene
Flat Bottom Sample Vial
Push on hinged lid
1.35mm
Push on hinged lid
Videos
Documentation