missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polyethylene Flat-Bottom Sample Vials

General purpose containers for laboratory weighing and storage needs.

Brand:  Kartell™ 735

Code : 02

Additional Details : Weight : 0.12596kg

 View more versions of this product

Product Code. 11301225

Quantity Price
1 £7.35 / Pack of 10
10 £6.73 / Pack of 10
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description

Description

  • Convenient push-on attached hinged lids
  • Lightweight and unbreakable
  • Translucent
  • Suitable for storing food stuff
Specifications

Specifications

35mL
Polyethylene
Flat Bottom Sample Vial
Push on hinged lid
1.6mm
Push on hinged lid
Videos
Documentation