Learn More
Sartorius Stedim Biotech™ Polyethersulfone (PES) Membrane Filters
General purpose membrane that provides excellent performance with most solutions when retentate recovery is of primary importance.
Brand: Sartorius Stedim Biotech™ 14668--63------D
244.83 GBP valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Additional Details : Weight : 0.01800kg
Description
- General purpose membrane that provides excellent performance with most solutions when retentate recovery is of primary importance.
- PES membranes exhibit no hydrophobic or hydrophilic interactions and are usually preferred for their low fouling characteristics, exceptional flux and broad pH range.
- Suitable for biopharma, research, buffer exchange, protein/DNA concentration
Specifications
MWCO, 10,000 Daltons | |
Biopharma, research, buffer exchange, protein/DNA concentration | |
10/pk. | |
64 mm |
PES | |
Ultrafilter | |
Ultrafilter | |
64 mm |