missing translation for 'onlineSavingsMsg'
Learn More

Synergy Healthcare Pouch flat peelable 75mm x 250mm

Brand:  Synergy Healthcare RF100200

Pouch flat peelable 75mm x 250mm

Code : 03

Additional Details : Weight : 0.25000kg

Product Code. 12798995

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.