missing translation for 'onlineSavingsMsg'
Learn More

Synergy Healthcare™ Azo™ Flat Pouches

High quality preformed, heat sealable, peelable sterile barrier packaging. Synergy Healthcare™ Azo™ Flat Pouches are suited to the sterilization of surgical instruments, laboratory equipment or small to medium sized sets using steam or Ethylene Oxide.

Brand:  Synergy Healthcare™ PF100250

Code : 03

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12340479

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Made from specially treated paper, less fibres released when pouch is opened and reducing the risk of contamination
  • New film technology increases impact strength of pouch film up to 10 times, virtually eliminates the shattering of film when opening
  • Indicators are placed between the 2 layers which make-up the pouch film, prevents offsetting and leakage of ink
  • Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications

Specifications

EN868-5, ISO11607-1, 93/42/EEC, ISO13485, ISO9001
250 x 100mm
Suitable for both steam and ethylene oxide sterilization
170 to 210°C
Cool dry place
135μm
Blue
Polyethylene
Heat seal
Yes
2400
No
6.30 kN/m
Yes
250mm
Width (Metric)
Videos
Documentation

Technical Documents