Learn More
0 Polyester/Polypropylene Pouch
Brand:  0 PF100300
Code : 03
Additional Details : Weight : 0.20000kg
Product Code. 12729005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Manufactured from transparent polyester/polypropylene film for strength and puncture resistance, and high quality wet strength MG kraft paper. Opens along width. Conforms to BS 6871.Suitable for both steam and ethylene oxide sterilisation.