missing translation for 'onlineSavingsMsg'
Learn More

Polyester acrylate/methacrylate, PEAM-1769, Thermo Scientific Chemicals

Catalog Number 15492667
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
921214-61-1
This item is not returnable. View return policy
921214-61-1

Recommended for use as a base resin or an additive to reduce stress

Recommended for use as a base resin or an additive to reduce stress

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Amber
Physical Form Amber liquid
Quantity 25 g
Chemical Name or Material Polyester acrylate/methacrylate, PEAM-1769

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'