missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410, Thermo Scientific Chemicals

Catalog Number 11315897
Click to view available options
Quantity:
50 mL
250 mL
1 L
This item is not returnable. View return policy
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
C[Si](C)(-*)O-*
This item is not returnable. View return policy
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
C[Si](C)(-*)O-*

Polydimethylsiloxane trimethylsiloxy terminated, is widely used as a surfactant, as a component of defoamers which are used to suppress the formation of foams. It is also used as a critical ingredient in water-repelling coatings. It is used as an active silicone fluid, as a stamp resin in the procedure of soft lithography, in optic telecommunications, in biomedical research, as elasticity of polymer networks, as adhesive materials and in skin-moisturizing lotions. It plays an important role in filler fluid in breast implants and as a sorbent for the analysis of headspace of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Boiling Point 85°C to 105°C (2 mmHg to 4 mmHg)
Melting Point -40°C
Density 0.88 g/mL
Solubility Information Slightly miscible with water. Immiscible with methanol and ethylene glycol.
Quantity 1 L
Flash Point 70°C (158°F)
Odor Odorless
Refractive Index 1.39
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'