missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 4000, Thermo Scientific Chemicals

Catalog Number 11309048
Click to view available options
Quantity:
50 mL
250 mL
1 L
This item is not returnable. View return policy
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
C[Si](C)(-*)O-*
This item is not returnable. View return policy
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
C[Si](C)(-*)O-*

Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppress the formation of foams and as a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Viscous Liquid
Boiling Point >225°C
Melting Point <-40°C
Density 0.960 g/mL
Solubility Information Slightly miscible with water. Immiscible with methanol and ethylene glycol.
Quantity 1 L
Flash Point 285°C (545°F)
Odor Odorless
Refractive Index 1.4015
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 4,000
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'