missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
4 kg
Description
Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Viscous Liquid |
Boiling Point | >205°C |
Melting Point | <-40°C |
Density | 0.977 g/mL |
Solubility Information | Not miscible with water. Insoluble in methanol,and ethylene glycol. |
Quantity | 50 g |
Flash Point | 321°C (610°F) |
Odor | Odorless |
Refractive Index | 1.4035 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'