missing translation for 'onlineSavingsMsg'
Learn More

Logistik Unicorp Laboratory coat WC20 108cm ladies stud cuffs 107cm length

Brand:  Logistik Unicorp 607-V3PE-108

Laboratory coat WC20 108cm ladies stud cuffs 107cm length

Code : 92

Additional Details : Weight : 1.00000kg

Product Code. 13152170

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.