missing translation for 'onlineSavingsMsg'
Learn More

Logistik Unicorp™ Polycotton Lab Coat

Brand:  Logistik Unicorp™ 607-V3PE-92

Code : 92

Additional Details : Weight : 1.00000kg

Product Code. 13111741

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Petrol blue
Polycotton
92cm
Lab Coat
stud
3
Ladies
107cm
Videos
Documentation

Technical Documents