Learn More
Kimberly-Clark™ polycarbonate safety eyewear.
Brand:  Kimberly-Clark™ 25652
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11725054
  • £45.35 / Pack of 12
Estimated Shipment
01-02-2018
Add to basket
Specifications
Specifications
Safety Eyewear
Providing workers and visitors with basic eye protection. Smoky eye for outdoor use, prevents eye fatigue due to glare
Polycarbonate
12/pk.
EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity.
0.264kg
Videos