Learn More
Masterflex™ Polycarbonate Manifolds
Regulate flow without leaking with these manifolds with luer locks. Cole Parmer™ Polycarbonate Manifolds feature polyurethane stems, HDPE handles and female ports and luer locks.
Brand: Masterflex™ 06464-85
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Purchase manifolds individually or in convenient packs of ten
- This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications
Specifications
180° | |
Luer Fitting | |
Polycarbonate | |
4 |