missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ Polycarbonate Isopore Media Membrane Filter

Brand:  Millipore™ TETP14250

Code : 71

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10657701

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

60L/min./cm2/psi
142mm
Plain
5 to 20%
140°C
For analyses of airborne contaminants & other particles using optical or electron microscopy
8μm
White
TETP
Screen filter
50Pack
18μm
Polycarbonate
Isopore Membrane Filter