missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Polyaniline, emeraldine salt from p-toluenesulfonic acid

Brand:  Alfa Aesar™ 043683.09

Code : NEW

Additional Details : CAS Number : 25233-30-1 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11370268

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Polyaniline
25233-30-1
Emeraldine salt from p-toluenesulfonic acid
MFCD00284320

Specifications

Polyaniline
25233-30-1
MFCD00284320
>300°C (dec.)
Emeraldine salt from p-toluenesulfonic acid
Powder
Dark green
10g
Videos
SDS