missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Polyaniline, emeraldine base

Conductive polymer

Brand:  Alfa Aesar™ 042581.14

Code : NEW

Additional Details : CAS Number : 25233-30-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11356119

  • £216.20 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Polyaniline
25233-30-1
Emeraldine base
MFCD00241446

Specifications

Polyaniline
25233-30-1
MFCD00241446
>200°C (dec.)
Emeraldine base
Powder
Dark brown
25g
Videos
SDS