missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinylbenzyl chloride), Thermo Scientific Chemicals

Catalog Number p-7058702
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
9080-67-5
−[CH2CH(C6H4CH2Cl-]n
MFCD00135445
This item is not returnable. View return policy
9080-67-5
−[CH2CH(C6H4CH2Cl-]n
MFCD00135445

In scientific and educational research. In synthesis of polymer supported reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Solid
Solubility Information Insoluble in water.
Quantity 2 g
Chemical Name or Material Poly (vinylbenzyl chloride)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'