missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl formal), Thermo Scientific Chemicals

Catalog Number p-7059214
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
9003-33-2
(C5H8O2)n
100.12
MFCD00084463
DFFZYNIDEOOVAU-UHFFFAOYSA-N
poly vinyl formal, ethenyloxy methoxy ethene, polyvinyl formal, ethene, 1,1'-methylenebis oxy bis-, homopolymer, vinyloxymethoxyethylene, methylene glycol divinyl ether, 1,1'-methylenebisoxy bisethene, ethene,1,1'-methylenebis oxy bis-9ci
165717
ethenoxymethoxyethene
*-CC1CC(-*)OCO1
This item is not returnable. View return policy
9003-33-2
(C5H8O2)n
100.12
MFCD00084463
DFFZYNIDEOOVAU-UHFFFAOYSA-N
poly vinyl formal, ethenyloxy methoxy ethene, polyvinyl formal, ethene, 1,1'-methylenebis oxy bis-, homopolymer, vinyloxymethoxyethylene, methylene glycol divinyl ether, 1,1'-methylenebisoxy bisethene, ethene,1,1'-methylenebis oxy bis-9ci
165717
ethenoxymethoxyethene
*-CC1CC(-*)OCO1

It is used in the anomalous sorption and pervaporation of aqueous organic mixtures by poly(vinyl acetal) membranes. Poly(vinyl formal) is mainly used in three general applications wire coatings, surface coatings and adhesives. The first and extensive application of poly(vinyl formal) is in the manufacture of a tough, adhesive and heat resistant wire enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Granular/ powder
Density 1.23 g/mL
Solubility Information Soluble in acetone, chloroform, dioxane, THF
Quantity 25 g
Chemical Name or Material Poly (vinyl formal)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'