missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals

Catalog Number p-7052138
Click to view available options
Quantity:
100 g
500 g
28210-41-5
MFCD00165973
4-styrenesulfonic acid, polystyrene sulfonate, tolevamer, styrene-4-sulphonic acid, 4-vinylbenzenesulfonic acid, benzenesulfonic acid, 4-ethenyl, poly 4-vinylbenzenesulfonic acid, tolevamer inn, poly styrene sulfonic acid sodium salt, unii-zsl2fb6gxn
28210-41-5
MFCD00165973
4-styrenesulfonic acid, polystyrene sulfonate, tolevamer, styrene-4-sulphonic acid, 4-vinylbenzenesulfonic acid, benzenesulfonic acid, 4-ethenyl, poly 4-vinylbenzenesulfonic acid, tolevamer inn, poly styrene sulfonic acid sodium salt, unii-zsl2fb6gxn

Poly(styrene sulfonic acid) is used in water softening and to remove hard ions like calcium and magnesium. In the medical field, it is used as potassium binders in acute and chronic kidney disease. Its sodium salt is used as a superplastifier in cement, as a dye improving agent for cotton, as proton exchange membranes in fuel cell applications. It acts as a solid acid catalyst in organic synthesis. It is an antistatic resin used for electrographic and electrophotographic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Amber
Density 1.1 g/mL
Quantity 100 g
UN Number UN2584
Solubility Information Miscible with water,ethanol,methanol,lower alcohols and glycols.
Concentration or Composition (by Analyte or Components) 30% w/v aq. soln.
Physical Form Liquid
Chemical Name or Material Poly (styrene sulfonic acid), M.W. 75,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'