missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh

For the preparation of functionalized beads used in solid-phase synthesis

Brand:  Alfa Aesar™ 041800.14

Code : NEW

Additional Details : CAS Number : 9003-70-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11318968

  • £90.20 / 25g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Poly(styrene-divinylbenzene)
9003-70-7
1% cross-linked
MFCD00146442

Specifications

Poly(styrene-divinylbenzene)
9003-70-7
MFCD00146442
Odorless
1% cross-linked
Beads
200 to 400 Mesh
25g
Videos
SDS