missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate), ∽ M.W. 15000 (GPC), Thermo Scientific Chemicals

Catalog Number p-186380
Click to view available options
Quantity:
50 g
100 g
Packaging:
Plastic bottle
9011-14-7
(C5H8O2)n
NaN
MFCD00134349
PMAMJWJDBDSDHV-UHFFFAOYSA-N
Methyl methacrylate, polymerized, PMMA
1-methoxy-2-methyl-1-oxopropan-2-yl
COC(=O)C(C)(-*)C-*
9011-14-7
(C5H8O2)n
NaN
MFCD00134349
PMAMJWJDBDSDHV-UHFFFAOYSA-N
Methyl methacrylate, polymerized, PMMA
1-methoxy-2-methyl-1-oxopropan-2-yl
COC(=O)C(C)(-*)C-*

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Beads
Linear Formula [-CH2C(CH3)(CO2CH3)-]n
Density 1.18
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf
Quantity 50 g
Flash Point >250°C
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Plastic bottle
Particle Size Bead size (typically): 10 to 100 micron
Chemical Name or Material Poly(methyl methacrylate), Average M.W. 15000 (GPC)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'