missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 4,600

Fluorosilicone lubricant

Brand:  Alfa Aesar™ 043229.06

Code : NEW

Additional Details : CAS Number : 63148-56-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11369308

  • £14.50 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Poly (methyl-3,3,3-trifluoropropylsiloxane)
M.W. 4,600
MFCD00241477
63148-56-1
[-Si(CH3)(CH3CH2CF3)O-]n

Specifications

Poly (methyl-3,3,3-trifluoropropylsiloxane)
63148-56-1
Liquid
MFCD00241477
Colorless
290°C (554°F)
M.W. 4,600
1.28g/mL
[-Si(CH3)(CH3CH2CF3)O-]n
1.382
5g
Videos
SDS