missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene oxide), ∽ M.W. 200,000, ACROS Organics™

Brand:  Acros Organics 178592500

Code : 29

Additional Details : CAS Number : 25322-68-3 Weight : 0.30000kg

Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10730881

  • £88.30 / 250g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

Poly(ethylene oxide)
25322-68-3
MFCD00081839
Approximate M.W. 200.000
(-CH2CH2O-)n
PEO

Specifications

Poly(ethylene oxide)
25322-68-3
(-CH2CH2O-)n
15, 7688
Authentic
Glass bottle
White to off-white
250g
Approximate M.W. 200.000
Granules or Crystalline Powder
MFCD00081839
PEO
1% max.
Solubility in water: soluble. Other solubilities: soluble in many organic solvents
65.0°C
Videos
SDS