missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene oxide), 100%, ∽ M.W. 900,000, Thermo Scientific Chemicals

Catalog Number p-186216
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEO
ethane-1,2-diol
[H]OCCO
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEO
ethane-1,2-diol
[H]OCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Melting Point 65.0°C to 67.0°C
Linear Formula (-CH2CH2O-)n
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Chemical Name or Material Poly(ethylene oxide), Approximate M.W. 900, 000
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'