missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene oxide), 100%, ∽ M.W. 900,000, ACROS Organics™

Brand:  Acros Organics 183221000

Code : 29

Additional Details : CAS Number : 25322-68-3 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10499660

  • £44.20 / 100g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Poly(ethylene oxide)
25322-68-3
MFCD00081839
Approximate M.W. 900, 000
(-CH2CH2O-)n
PEO

Specifications

Poly(ethylene oxide)
25322-68-3
(-CH2CH2O-)n
15, 7688
Authentic
Glass bottle
White
100g
Approximate M.W. 900, 000
Powder
MFCD00081839
PEO
1% max.
Solubility in water: soluble. Other solubilities: soluble in many organic solvents
65.0° to 67.0°C
Videos
SDS