missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene Glycol), Average M.W. 8000, ACROS Organics™
£41.87 - £275.04
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10005850
|
Acros Organics
418050010 |
1kg | Plastic Bottle |
£41.87
1kg |
|
||||
10065830
|
Acros Organics
418050030 |
3kg | Plastic drum |
£104.43
3kg |
|
||||
10309103
|
Acros Organics
418050100 |
10kg | Plastic drum |
£275.04
10kg |
|
||||
Chemical Identifiers
25322-68-3 | |
MFCD01779615 | |
PEG |
Specifications
Poly(ethylene glycol) | |
25322-68-3 | |
H(OCH2CH2)nOH | |
15, 7688 | |
12 to 16mg KOH/g | |
≤0.5% (K.F.) | |
55.0°C to 60.0°C | |
270°C |
Average M.W. 8000 | |
white flakes | |
MFCD01779615 | |
PEG | |
Plastic Bottle | |
300-450 mPa.s (20°C 50% in water) | |
1kg |