missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 600, Thermo Scientific Chemicals

Catalog Number p-186140
Click to view available options
Quantity:
1 L
5 L
Packaging:
Plastic bottle
Plastic drum
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Viscous Liquid After Melting
Melting Point 17.0°C to 22.0°C
Linear Formula H(OCH2CH2)nOH
Density 1.1265g/mL
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 1 L
Formula Weight 600
Flash Point 252°C
Hydroxyl Value 178 to 197mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.4660 to 1.4700
Specific Gravity 1.1265
Viscosity 16-19 mPa.s (20°C)
Water 0.5% max. (K.F.)
Chemical Name or Material Poly(ethylene glycol), Average M.W. 600
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'