missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene glycol), average M.W. 400, Thermo Scientific Chemicals
38.89 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Boiling Point | >200.0°C |
Melting Point | 4.0°C to 8.0°C |
Linear Formula | H(OCH2CH2)nOH |
Density | 1.1275g/mL |
Merck Index | 15, 7688 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in many organic solvents |
Quantity | 1 L |
Formula Weight | 400 |
Show More |
Safety and Handling
Product Identifier
- Poly(ethylene glycol), average M.W. 200
- Warning
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'