missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 300, Thermo Scientific Chemicals

Catalog Number p-3737757
51.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Clear liquid
Melting Point -15.0°C to +10.0°C
Linear Formula H(OCH2CH2)nOH
Density 1.1200g/mL
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 1 L
Flash Point >180°C
Hydroxyl Value 356 to 394mg KOH/g
Packaging Glass bottle
Refractive Index 1.4620 to 1.4650
Specific Gravity 1.12
Viscosity 85-100 mPa.s (20°C)
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less
compliance-icons
Product Identifier
  • Poly(ethylene glycol), average M.W. 200
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'