missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 200, Thermo Scientific Chemicals

Catalog Number p-3737988
Click to view available options
Quantity:
5 mL
1 L
5 L
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Melting Point -65.0°C
Linear Formula H(OCH2CH2)nOH
Density 1.1250g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 5 mL
Formula Weight 200
Flash Point 171°C
Hydroxyl Value 534 to 591mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.4580 to 1.4610
Specific Gravity 1.125
Viscosity 60-70 mPa.s (20°C)
Water 0.5% max. (K.F.)
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'