missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 1450, Thermo Scientific Chemicals

Catalog Number p-186087
Click to view available options
Quantity:
100 g
500 g
1 kg
3 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG
ethane-1,2-diol
[H]OCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Waxy Flakes
Melting Point 44.0°C to 48.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Quantity 100 g
Hydroxyl Value 70 to 80mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic bottle
Viscosity 36-42 mPa.s ( 20°C 50% in water)
Water 0.5% max. (K.F.)
Chemical Name or Material Poly(ethylene glycol), Average M.W. 1450
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'