missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene glycol), average M.W. 1000, Thermo Scientific Chemicals
29.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Waxy Solid |
Melting Point | 35.0°C to 40.0°C |
Linear Formula | H(OCH2CH2)nOH |
Merck Index | 15, 7688 |
Solubility Information | Solubility in water: 750g/L (20°C). Other solubilities: soluble in many organic solvents |
Quantity | 1 kg |
Formula Weight | 1000 |
Flash Point | >260°C |
Hydroxyl Value | 107 to 118mg KOH/g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'