missing translation for 'onlineSavingsMsg'
Learn More

Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.W. 19,000, ACROS Organics™

Brand:  Acros Organics 436200010

Code : 29

Additional Details : CAS Number : 26780-50-7 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 11936981

  • £49.40 / 1g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Poly(DL-lactide-co-glycolide)
26780-50-7

Specifications

Poly(DL-lactide-co-glycolide)
Crystalline Powder or Granules
45 to 55% (Lactide), 45 to 55% (Glycolide)
0.15 to 0.25 dL/g
1g
26780-50-7
Glass bottle
Solubility in water: soluble.
White to tan
Videos
SDS